siRNA Details
| Gene Symbol: RGD1304884 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107436.1 | NP_001100906.1 | 8 | 1135 | ||
| XM_006255743.2 | XP_006255805.1 | 7 | 983 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1304884| Gene Symbol: RGD1304884 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107436.1 | NP_001100906.1 | 8 | 1135 | ||
| XM_006255743.2 | XP_006255805.1 | 7 | 983 | |||