siRNA Details
| Gene Symbol: Gse1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006255748.2 | XP_006255810.1 | 13, 14 | 3379 | ||
| XM_008772656.1 | XP_008770878.1 | 12, 13 | 9439 | |||
| XM_008772657.1 | XP_008770879.1 | 11, 12 | 9322 | |||
| XM_008772658.1 | XP_008770880.1 | 14, 15 | 3349 | |||
| XM_008772659.1 | XP_008770881.1 | 13, 14 | 4707 | |||
| XM_008772660.1 | XP_008770882.1 | 12, 13 | 3160 | |||
| XM_008772661.1 | XP_008770883.1 | 12, 13 | 3754 | |||
| XM_008772662.1 | XP_008770884.1 | 12, 13 | 4487 | |||
| XM_226533.8 | XP_226533.5 | 13, 14 | 3973 | |||

Gse1
Genomic Map