siRNA Details
| Gene Symbol: Gse1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006255748.2 | XP_006255810.1 | 9 | 2267 | ||
| XM_008772656.1 | XP_008770878.1 | 8 | 8327 | |||
| XM_008772657.1 | XP_008770879.1 | 8 | 8327 | |||
| XM_008772658.1 | XP_008770880.1 | 10 | 2237 | |||
| XM_008772659.1 | XP_008770881.1 | 9 | 3595 | |||
| XM_008772660.1 | XP_008770882.1 | 8 | 2048 | |||
| XM_008772661.1 | XP_008770883.1 | 8 | 2642 | |||
| XM_008772662.1 | XP_008770884.1 | 8 | 3375 | |||
| XM_226533.8 | XP_226533.5 | 9 | 2861 | |||

Gse1
Genomic Map