siRNA Details
| Gene Symbol: Snai3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107439.1 | NP_001100909.1 | 1, 2 | 143 | ||
| XM_008772612.1 | XP_008770834.1 | 2, 3 | 4810 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Snai3| Gene Symbol: Snai3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107439.1 | NP_001100909.1 | 1, 2 | 143 | ||
| XM_008772612.1 | XP_008770834.1 | 2, 3 | 4810 | |||