siRNA Details
Gene Symbol: Trim67 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135715.1 | NP_001129187.1 | 6 | 1981 | ||
XM_006255815.2 | XP_006255877.1 | 6 | 2272 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Trim67 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135715.1 | NP_001129187.1 | 6 | 1981 | ||
XM_006255815.2 | XP_006255877.1 | 6 | 2272 |