siRNA Details
| Gene Symbol: Disc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175596.2 | NP_783186.2 | 12 | 2323 | ||
| XM_008772616.1 | XP_008770838.1 | 12 | 3718 | |||
| GenBank mRNA | AY177674.2 | - | 12 | 2323 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Disc1| Gene Symbol: Disc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175596.2 | NP_783186.2 | 12 | 2323 | ||
| XM_008772616.1 | XP_008770838.1 | 12 | 3718 | |||
| GenBank mRNA | AY177674.2 | - | 12 | 2323 | ||