siRNA Details
Gene Symbol: Disc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175596.2 | NP_783186.2 | 6 | 1440 | ||
XM_008772616.1 | XP_008770838.1 | 6 | 2835 | |||
GenBank mRNA | AY177674.2 | - | 6 | 1440 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Disc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175596.2 | NP_783186.2 | 6 | 1440 | ||
XM_008772616.1 | XP_008770838.1 | 6 | 2835 | |||
GenBank mRNA | AY177674.2 | - | 6 | 1440 |