siRNA Details
| Gene Symbol: Pcnxl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006255821.2 | XP_006255883.1 | 28, 29 | 5809 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pcnxl2| Gene Symbol: Pcnxl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006255821.2 | XP_006255883.1 | 28, 29 | 5809 | ||