siRNA Details
Gene Symbol: Def8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024774.1 | NP_001019945.1 | 8 | 898 | ||
XM_006255769.2 | XP_006255831.1 | 8 | 985 | |||
XM_008772618.1 | XP_008770840.1 | 8 | 974 | |||
GenBank mRNA | BC097376.1 | - | 8 | 898 | 7444965 |
