siRNA Details
| Gene Symbol: Def8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024774.1 | NP_001019945.1 | 5, 6 | 589 | ||
| XM_006255769.2 | XP_006255831.1 | 5, 6 | 676 | |||
| XM_008772618.1 | XP_008770840.1 | 5, 6 | 665 | |||
| GenBank mRNA | BC097376.1 | - | 5, 6 | 589 | 7444965 | |

Def8
Genomic Map