siRNA Details
| Gene Symbol: Nhlrc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107444.1 | NP_001100914.1 | 2 | 664 | ||
| XM_006231645.2 | XP_006231707.1 | 2 | 664 | |||
| XM_008760536.1 | XP_008758758.1 | 2 | 438 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nhlrc2| Gene Symbol: Nhlrc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107444.1 | NP_001100914.1 | 2 | 664 | ||
| XM_006231645.2 | XP_006231707.1 | 2 | 664 | |||
| XM_008760536.1 | XP_008758758.1 | 2 | 438 | |||