siRNA Details
| Gene Symbol: Fam45a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127681.1 | NP_001121153.1 | 7 | 854 | ||
| XM_006231682.1 | XP_006231744.1 | 7 | 846 | |||
| GenBank mRNA | BC167081.1 | - | 7 | 823 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam45a| Gene Symbol: Fam45a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127681.1 | NP_001121153.1 | 7 | 854 | ||
| XM_006231682.1 | XP_006231744.1 | 7 | 846 | |||
| GenBank mRNA | BC167081.1 | - | 7 | 823 | ||