siRNA Details
| Gene Symbol: Clca1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107449.1 | NP_001100919.1 | 14 | 2634 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Clca1| Gene Symbol: Clca1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107449.1 | NP_001100919.1 | 14 | 2634 | ||