siRNA Details
| Gene Symbol: Odf2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006233440.2 | XP_006233502.1 | 2 | 665 | ||
| XM_006233442.2 | XP_006233504.1 | 1 | 6505 | |||
| XM_006233444.2 | XP_006233506.1 | 1 | 6504 | |||
| XM_006233446.2 | XP_006233508.1 | 1 | 6505 | |||
| XM_006233447.2 | XP_006233509.1 | 1 | 6503 | |||
| XM_006233448.2 | XP_006233510.1 | 1 | 6503 | |||
| XM_006233449.2 | XP_006233511.1 | 1 | 6503 | |||
| XM_008761524.1 | XP_008759746.1 | 1 | 6505 | |||
| XM_008761525.1 | XP_008759747.1 | 2 | 793 | |||

Odf2l
Genomic Map