siRNA Details
| Gene Symbol: Syde2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749414.3 | XP_003749462.1 | 3 | 3503 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Syde2| Gene Symbol: Syde2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749414.3 | XP_003749462.1 | 3 | 3503 | ||