siRNA Details
| Gene Symbol: Rnf146 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012060.2 | NP_001012060.2 | 3 | 1197 | ||
| XM_006227744.2 | XP_006227806.1 | 3 | 1254 | |||
| XM_006227745.2 | XP_006227807.1 | 4 | 1302 | |||
| GenBank mRNA | BC083675.1 | - | 3 | 1197 | 7135728 | |

Rnf146
Genomic Map