siRNA Details
| Gene Symbol: Brd9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107453.1 | NP_001100923.1 | 7, 8 | 883 | ||
| XM_006227785.2 | XP_006227847.1 | 7, 8 | 1228 | |||
| XM_006227786.2 | XP_006227848.1 | 7, 8 | 1168 | |||
| XM_006227787.2 | XP_006227849.1 | 7, 8 | 1165 | |||

Brd9
Genomic Map