siRNA Details
| Gene Symbol: Slc12a7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013144.2 | NP_001013162.2 | 8, 9 | 1183 | ||
| XM_006227789.2 | XP_006227851.1 | 8, 9 | 1308 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc12a7| Gene Symbol: Slc12a7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013144.2 | NP_001013162.2 | 8, 9 | 1183 | ||
| XM_006227789.2 | XP_006227851.1 | 8, 9 | 1308 | |||