siRNA Details
Gene Symbol: Map3k4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107456.1 | NP_001100926.1 | 12 | 3090 | ||
XM_006227872.2 | XP_006227934.1 | 14 | 3485 | |||
XM_006227873.2 | XP_006227935.1 | 14 | 3485 | |||
XM_006227874.2 | XP_006227936.1 | 13 | 3242 |
