siRNA Details
| Gene Symbol: Ccr6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013145.1 | NP_001013163.1 | 2 | 1004 | ||
| XM_006227922.2 | XP_006227984.1 | 3 | 1114 | |||
| XM_008758755.1 | XP_008756977.1 | 2 | 881 | |||
| XM_008758756.1 | XP_008756978.1 | 2 | 2930 | |||
| GenBank mRNA | BC091197.1 | - | 2 | 1004 | 7374663 | |
| EF547659.1 | - | 1 | 739 | |||
| FQ225325.1 | - | 1 | 377 | |||
| FQ225692.1 | - | 2 | 881 | |||
| FQ226017.1 | - | 2 | 881 | |||
| FQ227530.1 | - | 1 | 731 | |||

Ccr6
Genomic Map