siRNA Details
| Gene Symbol: Chd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107465.1 | NP_001100935.1 | 15 | 2444 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_007690, L10410, AK140386, AK015218, AK049686, and others.

Chd1
Genomic Map