siRNA Details
| Gene Symbol: Fam120b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107466.3 | NP_001100936.2 | 2 | 1323 | ||
| XM_006231141.2 | XP_006231203.1 | 2 | 1452 | |||
| XM_006231142.2 | XP_006231204.1 | 2 | 1451 | |||
| XM_008758761.1 | XP_008756983.1 | 2 | 1164 | |||
| GenBank mRNA | FQ210918.1 | - | 2 | 1323 | ||

Fam120b
Genomic Map