siRNA Details
| Gene Symbol: Lats1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134543.2 | NP_001128015.2 | 8 | 3506 | ||
| XM_006227614.2 | XP_006227676.1 | 7 | 4148 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lats1| Gene Symbol: Lats1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134543.2 | NP_001128015.2 | 8 | 3506 | ||
| XM_006227614.2 | XP_006227676.1 | 7 | 4148 | |||