siRNA Details
| Gene Symbol: Fbxo30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007690.1 | NP_001007691.1 | 2 | 1608 | ||
| XM_006227640.2 | XP_006227702.1 | 2 | 1650 | |||
| GenBank mRNA | BC083823.1 | - | 2 | 1608 | 7111792 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fbxo30| Gene Symbol: Fbxo30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007690.1 | NP_001007691.1 | 2 | 1608 | ||
| XM_006227640.2 | XP_006227702.1 | 2 | 1650 | |||
| GenBank mRNA | BC083823.1 | - | 2 | 1608 | 7111792 | |