siRNA Details
| Gene Symbol: Mboat7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134978.1 | NP_001128450.1 | 465 | |||
| GenBank mRNA | BC169089.1 | - | 465 | 5598892 | ||
| Gene Symbol: LOC103689982 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008758837.1 | XP_008757059.1 | 4 | 465 | ||

Mboat7
Genomic Map