siRNA Details
| Gene Symbol: RGD1564236 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006245569.2 | XP_006245631.1 | 2 | 1825 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1564236| Gene Symbol: RGD1564236 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006245569.2 | XP_006245631.1 | 2 | 1825 | ||