siRNA Details
| Gene Symbol: Ssc5d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134545.1 | NP_001128017.1 | 14 | 3873 | ||
| XM_006228257.1 | XP_006228319.1 | 13 | 4304 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ssc5d| Gene Symbol: Ssc5d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134545.1 | NP_001128017.1 | 14 | 3873 | ||
| XM_006228257.1 | XP_006228319.1 | 13 | 4304 | |||