siRNA Details
| Gene Symbol: Dnaaf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271115.1 | NP_001258044.1 | 10, 11 | 1220 | ||
| XM_008758882.1 | XP_008757104.1 | 10, 11 | 1409 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dnaaf3| Gene Symbol: Dnaaf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271115.1 | NP_001258044.1 | 10, 11 | 1220 | ||
| XM_008758882.1 | XP_008757104.1 | 10, 11 | 1409 | |||