siRNA Details
Gene Symbol: Dnaaf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271115.1 | NP_001258044.1 | 10 | 1158 | ||
XM_008758882.1 | XP_008757104.1 | 10 | 1347 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Dnaaf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271115.1 | NP_001258044.1 | 10 | 1158 | ||
XM_008758882.1 | XP_008757104.1 | 10 | 1347 |