siRNA Details
| Gene Symbol: LOC102552463 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008758848.1 | XP_008757070.1 | 3 | 675 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102552463| Gene Symbol: LOC102552463 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008758848.1 | XP_008757070.1 | 3 | 675 | ||