siRNA Details
Gene Symbol: Sae1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012063.1 | NP_001012063.1 | 3 | 276 | ||
GenBank mRNA | BC079411.1 | - | 3 | 276 | 7113315 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Sae1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012063.1 | NP_001012063.1 | 3 | 276 | ||
GenBank mRNA | BC079411.1 | - | 3 | 276 | 7113315 |