siRNA Details
| Gene Symbol: Cic | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107490.3 | 15 | 3604 | |||
| XM_008758933.1 | XP_008757155.1 | 16 | 7121 | |||
| XM_008758934.1 | XP_008757156.1 | 16 | 7385 | |||
| XM_008758935.1 | XP_008757157.1 | 16 | 7118 | |||
| XM_008758936.1 | XP_008757158.1 | 16 | 7121 | |||
| XM_008758937.1 | XP_008757159.1 | 16 | 7118 | |||
| XM_008758938.1 | XP_008757160.1 | 15 | 3803 | |||
| XM_008758939.1 | XP_008757161.1 | 15 | 3801 | |||
| XM_008758940.1 | XP_008757162.1 | 15 | 3798 | |||
| GenBank mRNA | CB700599.1 | - | 2 | 249 | ||
| EV777117.1 | - | 1 | 154 | |||

Cic
Genomic Map