siRNA Details
| Gene Symbol: Cic | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107490.3 | 9 | 1389 | |||
| XM_008758933.1 | XP_008757155.1 | 10 | 4900 | |||
| XM_008758934.1 | XP_008757156.1 | 10 | 5164 | |||
| XM_008758935.1 | XP_008757157.1 | 10 | 4900 | |||
| XM_008758936.1 | XP_008757158.1 | 10 | 4900 | |||
| XM_008758937.1 | XP_008757159.1 | 10 | 4900 | |||
| XM_008758938.1 | XP_008757160.1 | 9 | 1582 | |||
| XM_008758939.1 | XP_008757161.1 | 9 | 1583 | |||
| XM_008758940.1 | XP_008757162.1 | 9 | 1583 | |||
| GenBank mRNA | CB811515.1 | - | 2 | 142 | ||

Cic
Genomic Map