siRNA Details
| Gene Symbol: Sptbn4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100845.2 | NP_001094315.2 | 17, 18 | 3976 | ||
| XM_008759140.1 | XP_008757362.1 | 17, 18 | 4017 | |||
| XM_008759141.1 | XP_008757363.1 | 17, 18 | 4017 | |||

Sptbn4
Genomic Map