siRNA Details
| Gene Symbol: Sptbn4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100845.2 | NP_001094315.2 | 3 | 344 | ||
| XM_008759140.1 | XP_008757362.1 | 3 | 385 | |||
| XM_008759141.1 | XP_008757363.1 | 3 | 385 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sptbn4| Gene Symbol: Sptbn4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100845.2 | NP_001094315.2 | 3 | 344 | ||
| XM_008759140.1 | XP_008757362.1 | 3 | 385 | |||
| XM_008759141.1 | XP_008757363.1 | 3 | 385 | |||