siRNA Details
Gene Symbol: Dyrk1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107496.1 | NP_001100966.1 | 6 | 802 | ||
XM_006228621.2 | XP_006228683.1 | 6 | 966 | |||
XM_006228622.2 | XP_006228684.1 | 6 | 966 | |||
XM_006228623.2 | XP_006228685.1 | 6 | 966 | |||
XM_006228624.2 | XP_006228686.1 | 6 | 817 |
