siRNA Details
| Gene Symbol: Nfkbid | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003748825.3 | XP_003748873.1 | 4 | 608 | ||
| XM_006228804.2 | XP_006228866.1 | 3 | 1094 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nfkbid| Gene Symbol: Nfkbid | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003748825.3 | XP_003748873.1 | 4 | 608 | ||
| XM_006228804.2 | XP_006228866.1 | 3 | 1094 | |||