siRNA Details
| Gene Symbol: Uri1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107507.1 | NP_001100977.1 | 3 | 391 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Uri1| Gene Symbol: Uri1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107507.1 | NP_001100977.1 | 3 | 391 | ||