siRNA Details
| Gene Symbol: Myh14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100690.1 | NP_001094160.1 | 11, 12 | 1240 | ||
| XM_006229056.2 | XP_006229118.1 | 12, 13 | 1440 | |||
| XM_006229057.2 | XP_006229119.1 | 11, 12 | 1416 | |||

Myh14
Genomic Map