siRNA Details
| Gene Symbol: Pnkp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004259.1 | NP_001004259.1 | 4, 5 | 635 | ||
| XM_008759371.1 | XP_008757593.1 | 3, 4 | 1017 | |||
| GenBank mRNA | BC079307.1 | - | 4, 5 | 635 | 7134659 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pnkp| Gene Symbol: Pnkp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004259.1 | NP_001004259.1 | 4, 5 | 635 | ||
| XM_008759371.1 | XP_008757593.1 | 3, 4 | 1017 | |||
| GenBank mRNA | BC079307.1 | - | 4, 5 | 635 | 7134659 | |