siRNA Details
| Gene Symbol: Pnkp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004259.1 | NP_001004259.1 | 1, 2 | 218 | ||
| XM_006229058.2 | XP_006229120.1 | 1, 2 | 650 | |||
| GenBank mRNA | BC079307.1 | - | 1, 2 | 218 | 7134659 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pnkp| Gene Symbol: Pnkp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004259.1 | NP_001004259.1 | 1, 2 | 218 | ||
| XM_006229058.2 | XP_006229120.1 | 1, 2 | 650 | |||
| GenBank mRNA | BC079307.1 | - | 1, 2 | 218 | 7134659 | |