siRNA Details
| Gene Symbol: Cpt1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034925.2 | NP_001030097.2 | 18 | 2345 | ||
| XM_006229066.2 | XP_006229128.1 | 18 | 3367 | |||
| XM_008759376.1 | XP_008757598.1 | 17 | 3511 | |||
| XM_008759377.1 | XP_008757599.1 | 16 | 2348 | |||
| GenBank mRNA | BC105882.1 | - | 9 | 1108 | ||

Cpt1c
Genomic Map