siRNA Details
| Gene Symbol: Plekha4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199101.1 | NP_954532.1 | 3 | 442 | ||
| XM_006229071.1 | XP_006229133.1 | 3 | 598 | |||
| XM_006229072.1 | XP_006229134.1 | 3 | 598 | |||
| GenBank mRNA | BC061738.1 | - | 3 | 442 | 5599064 | |

Plekha4
Genomic Map