siRNA Details
| Gene Symbol: Ntn5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001173526.1 | NP_001166997.1 | 3 | 745 | ||
| XM_008759380.1 | XP_008757602.1 | 5 | 2055 | |||
| XM_008759381.1 | XP_008757603.1 | 6 | 1905 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ntn5| Gene Symbol: Ntn5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001173526.1 | NP_001166997.1 | 3 | 745 | ||
| XM_008759380.1 | XP_008757602.1 | 5 | 2055 | |||
| XM_008759381.1 | XP_008757603.1 | 6 | 1905 | |||