siRNA Details
Gene Symbol: Ano5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006229266.2 | XP_006229328.1 | 20 | 3625 | ||
XM_006229268.2 | XP_006229330.1 | 20 | 3516 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ano5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006229266.2 | XP_006229328.1 | 20 | 3625 | ||
XM_006229268.2 | XP_006229330.1 | 20 | 3516 |