siRNA Details
| Gene Symbol: Ano5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006229266.2 | XP_006229328.1 | 1 | 849 | ||
| XM_006229268.2 | XP_006229330.1 | 1 | 848 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ano5| Gene Symbol: Ano5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006229266.2 | XP_006229328.1 | 1 | 849 | ||
| XM_006229268.2 | XP_006229330.1 | 1 | 848 | |||