siRNA Details
| Gene Symbol: Zmat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_002730284.4 | XP_002730330.1 | 7 | 1697 | ||
| XM_006257246.2 | XP_006257308.1 | 6 | 1446 | |||
| XM_006257247.1 | XP_006257309.1 | 8 | 772 | |||
| XM_006257248.1 | XP_006257310.1 | 9 | 853 | |||
| XM_006257250.2 | XP_006257312.1 | 8 | 727 | |||
| XM_008773457.1 | XP_008771679.1 | 7 | 4106 | |||
| XM_008773458.1 | XP_008771680.1 | 7 | 2838 | |||

Zmat1
Genomic Map