siRNA Details
| Gene Symbol: Synm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134858.3 | NP_001128330.1 | 4 | 3313 | ||
| XM_006229337.2 | XP_006229399.1 | 4 | 3308 | |||
| GenBank mRNA | AB091769.1 | - | 4 | 3221 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Synm| Gene Symbol: Synm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134858.3 | NP_001128330.1 | 4 | 3313 | ||
| XM_006229337.2 | XP_006229399.1 | 4 | 3308 | |||
| GenBank mRNA | AB091769.1 | - | 4 | 3221 | ||