siRNA Details
| Gene Symbol: Unc45a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037647.1 | NP_001032736.1 | 12, 13 | 1870 | ||
| XM_006229432.2 | XP_006229494.1 | 12, 13 | 1821 | |||
| GenBank mRNA | BC107919.1 | - | 12, 13 | 1870 | 7457637 | |

Unc45a
Genomic Map