siRNA Details
| Gene Symbol: Pde8a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198767.1 | NP_942062.1 | 3 | 322 | ||
| XM_006229438.2 | XP_006229500.1 | 3 | 509 | |||
| XM_006229439.2 | XP_006229501.1 | 3 | 521 | |||
| GenBank mRNA | AB092696.1 | - | 3 | 322 | ||

Pde8a
Genomic Map